missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™-overall met capuchon
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ 600 Plus with Socks is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
13.50 - 995.00
Specificatie
Producttype | Overall met capuchon |
---|---|
Materiaal | Tyvek™ stof |
Voor eenmalig gebruik | Single-use |
Soort taille | Elastisch |
Ankle Style | Elastisch, Attached Socks, Boot Overflaps |
Productcode | Merk | Maat | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Productcode | Merk | Maat | Prijs | Hoeveelheid en beschikbaarheid | |||||
11703073
|
DuPont™
D13398935 |
X-Large |
13.50
1 stuk |
![]() |
|||||
12322259
|
DuPont™
D13398935 |
X-Large |
256.00
|
|
|||||
11843510
|
DuPont™
D13398935 |
X-Large |
969.00
80 stuks |
|
|||||
11833520
|
DuPont™
D13495777 |
Small |
969.00
|
![]() |
|||||
11773063
|
DuPont™
D13495777 |
Small |
13.50
1 stuk |
![]() |
|||||
12392249
|
DuPont™
D13495777 |
Small |
248.00
|
![]() |
|||||
11843520
|
DuPont™
D13495765 |
Medium |
969.00
|
|
|||||
12302259
|
DuPont™
D13495765 |
Medium |
248.00
|
|
|||||
11793063
|
DuPont™
D13398948 |
Large |
13.50
|
![]() |
|||||
12312259
|
DuPont™
D13398948 |
Large |
256.00
20 stuks |
|
|||||
11833510
|
DuPont™
D13398948 |
Large |
969.00
|
![]() |
|||||
11823520
|
DuPont™
D13398927 |
2X-Large |
969.00
|
|
|||||
11713073
|
DuPont™
D13398927 |
2X-Large |
13.50
|
![]() |
|||||
12332259
|
DuPont™
D13398927 |
2X-Large |
256.00
20 stuks |
![]() |
|||||
12342259
|
DuPont™
D13495744 |
3X-Large |
254.00
20 stuks |
![]() |
|||||
12623700
|
DuPont™
D13495744 |
3X-Large |
995.00
80 stuks |
|
|||||
Beschrijving
- Hooded coverall with integrated socks and additional knee-length boot flap.
- Available in white in sizes S to XXXL.
- Robust yet lightweight (<250g per garment).
- Self-adhesive chin flap for tight seal of suit to the mask.
- Elasticated face, wrists and ankles as well as glued-in waist elastic.
- Elasticated thumb loops prevent sleeves from riding up.
Specificatie
Overall met capuchon | |
Single-use | |
Elastisch, Attached Socks, Boot Overflaps | |
Hooded | |
Zipper with Self-adhesive Flap and Chin Flap | |
Categorie III, type 4B, 5 en 6, EN 1073-2 EN 14126 | |
(EU) 2016/425, EN 14126, EN 1149-5 | |
80/Pk. |
Tyvek™ stof | |
Elastisch | |
Thumb Loop | |
Elastic | |
Gestikt en afgeplakt, Taped | |
Self-adhesive chin flap | |
For maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or in research and biosecurity laboratories, medical applications and when exposed to biological hazards |
The user should make the risk assessment to determine the protection required.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title