missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 500 Lab Coats
Tyvek™ Lab coats offer enhanced protection for body parts that are more exposed to hazardous substances.
373.00
Specificatie
Producttype | Chemical Resistant Lab Coat |
---|---|
Geslacht | Unisex |
Kleur | White |
Type sluiting | Zipper |
Soort manchet | Elastic |
Beschrijving
- Composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
- Permeable to both air and water vapour
- Offers an excellent barrier against fine particles and fibres (down to 1 micron in size)
- Certified according to Regulation (EU) 2016/425
- Partial body chemical protective clothing, Category III, Type PB (6-B)
- EN 14126 (barrier to infective agents)
- Antistatic treatment (EN 1149-5) on both sides
- Applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting, and general clean-up
Specificatie
Chemical Resistant Lab Coat | |
White | |
Elastic | |
Non-sterile | |
Stitched |
Unisex | |
Zipper | |
Limited-use | |
Chemicals |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title