Learn More
DuPont™ Tyvek™ 500 Boot Covers
Offers enhanced protection for body parts that are more exposed to hazardous substances. Tyvek™ 500 Boot Covers are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
275.00 - 326.00
Specificatie
Maat Eén maat
Materiaal HDPE
Kleur Wit
Voor eenmalig gebruik Single-use
Type sluiting Elastic with Ties
Meer specificaties weergeven
Producten 2
Productcode Merk Zooltype Prijs Hoeveelheid en beschikbaarheid  
Productcode Merk Zooltype Prijs Hoeveelheid en beschikbaarheid  
11763083
Bekijk documenten
DuPont™
D13395724
Standard
275.00
200 stuks
Only null left
Toevoegen aan winkelmandje
11803510
Bekijk documenten
DuPont™
D13395989
Anti-slip
326.00
Only null left
Edge
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Knee-length overboot available in white.
  • Fixation ties.
  • Available in one size.
  • With elasticated top and stitched internal seams.
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols.
  • Excellent barrier against fine particles and fibers (down to 1 micron in size).
  • Ultra-low-linting and antistatically treated.
  • Available with standard or slip-retardant sole.
  • Applications include:
    • Pharmaceutical Handling
    • Chemical Protection
    • Lead and Asbestos Abatement/Remediation
    • General Maintenance/Operations
    • Spray Painting
    • General Clean-up
Specificatie
Specificatie
Eén maat
Wit
Elastic with Ties
200 Pk.
480 mm
Overschoen
HDPE
Single-use
PPE Category I, EN 1149-5
18.9 in.
Fluid
Video's
SDS
Documentatie
Documentatie
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.