Learn More
Thermo Scientific™ TSX Series High-Performance Undercounter Lab Refrigerators



For a better environment inside and out - Introducing our new undercounter lab refrigerator.
Brand: Thermo Scientific™ TSX505SA
4460.14 EUR valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2° to 8°C. The Series' V-drive is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Unique to the TSX505, our dual refrigeration solution operates through the action of two synchronized solid-state heat pumps. The two solid-state heat pumps work to cool a combination of cold wall and forced air to help ensure that conditions are optimal for the most sensitive applications
Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
Designed to maintain temperature uniformity to protect medical, pharmaceutical-grade products, laboratory media, reagents and more.- Unique V-drive technology is designed to save energy without compromising the protection of stored products
- U.S. Environmental Protection Agency SNAP (Significant New Alternatives Policy) compliant refrigerants
- Quiet operation at just 35dbA (43 during pull down)
- Reduced energy consumption
- Sleek exterior design and touch screen
- Bright, easy-to-adjust interior LED lighting
- Touchscreen controller with password protection
- Datalogging with USB download functionality
- Battery backup
- Choice of reversible glass or solid door
- Air or glycol temperature display
- Automated locking
- 1" rear access port
- Remote alarm contacts
- Audible and visual alarms: power fail, high temp, low temp, low battery, memory full, door ajar
- ADA compliant
- Compatible with most major electronic locks*
*Data on file.

Specifications
TSX Series High-Performance Undercounter Refrigerator, Solid Door | |
5.5 cu. ft. | |
3°C to 7°C | |
Single Solid | |
3 | |
Undercounter | |
TSX Series | |
Alarm | |
20.5 x 19.5 x 23 in.(52 x 49.5 x 58.4 cm) | |
26 in. | |
66 cm | |
31.8 in. | |
79.5 cm | |
23.6 in. | |
60 cm | |
10 A Max / 3 A Steady State | |
100/240 V | |
50/60 Hz | |
155 lb. | |
119 lb. | |
45 Boxes per Level (9 x 5 Pattern), 225 Boxes per Shelf (5 Levels per Shelf, 2 Shelves per Unit), +45 Boxes (1 Additional Level Above Top Shelf) |
High-Performance Refrigerator | |
156 L | |
R744 | |
1 | |
CE, REACH, RoHS, UL | |
Full color TFT-LCD; 4.3”; 480 x 272 | |
Automatic | |
Smart-Vue Compatible | |
26 x 23.6 x 31.8 in. (66 x 60 x 81 cm) | |
20.5 in. | |
52 cm | |
23 in. | |
58.4 cm | |
19.5 in. | |
49.5 cm | |
NEMA 5-15 P | |
100 - 240 V 50-60 Hz | |
Energy efficient | |
71 kg | |
53.98 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.