missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG General Purpose Refrigerators - Crated
GREENER_CHOICE

Product Code. p-7217284 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
400 L
700 L
1500 L
Door Style:
Glass
Solid
Plug Type:
EU
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Door Style Plug Type unitSize
17167748 400 L Glass EU Each
17107758 400 L Solid EU Each
17187758 700 L Glass EU Each
17127768 700 L Solid EU Each
17147738 1500 L Glass EU Each
17187738 1500 L Solid EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17167748 Supplier Thermo Scientific™ Supplier No. TSG400REGCV

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The TSG Series of general purpose refrigerators are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.

The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from 400L to 1500L double door
  • Glass door (triple pane) and solid door configurations
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY

Specifications

Description TSG Series General Purpose Laboratory Refrigerator
Capacity (Metric) 400 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) +5°C
Refrigerant R290
Door Style Glass
No. of Doors 1
Shelves 5 Adjustable
Capacity (Metric) Shelf 30 kg
Cabinet Material Steel
Exterior Finish Painted Steel
Interior Painted Steel
Certifications/Compliance CE
Casters Yes
Control Type Digital Microprocessor
Ports 1 Rear Access Port
Warranty 12 months
Defrost Adaptive Defrost
Packaging Type Crated
Monitoring Options Alarm
Access Security Key Lock
Smart View Compatibility Smart-Vue Pro Compatible
Dimensions (D x W x H) Interior 493 x 500 x 1405 mm
Dimensions (L x W x H) Exterior 655 x 600 x 1955 mm
Plug Type EU
Voltage 230 V
Frequency 50 Hz
Green Features Energy Efficient
Shipping Weight (Metric) 150 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.