missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Reagent Grade, 99%, Honeywell™

Product Code. 15678190 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Ampule
1 L
10 Ampules
100 mL
2 L
25 mL
5 mL
5 x 10 Ampules
500 mL
Unit Size:
1 L
1 ampoule
10 ampoules
100mL
2 L
25mL
500mL
5mL
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15678190 10 Ampules 10 ampoules
15698490 5 mL 5mL
15688490 25 mL 25mL
15668190 100 mL 100mL
15628200 500 mL 500mL
15608200 1 L 1 L
15618200 2 L 2 L
15698190 1 Ampule 1 ampoule
15638200 5 x 10 Ampules Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15678190 Supplier Honeywell Research Chemicals Supplier No. T650810AMP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Spécification

Linear Formula CF3COOH
UN Number UN2699
Formula Weight 114.02
Percent Purity 99%
Density 1.489g/mL at 20°C
Quantity 10 Ampules
Flash Point >100°C (>212°F)
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid
compliance-icons compliance-icons
Identifiant produit
  • Trifluoroacetic acid
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Mention de danger
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Conseils de prudence
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.