missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid-d, 99.5%(Isotopic)

Product Code. 11324468
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 599-00-8 | C2HF3O2 | 115.029 g/mol

Trifluoroacetic acid-d is employed as solvent to investigate the conformations of cyclic peptides by proton NMR spectra. It is also used to compose NMR solvent mixture to investigate the NMR spectra of high molecular weight polyesters such as polyglycolide (PG) and poly(ethylene terephthalate)and as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.029
MDL Number MFCD00044563
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
Synonym trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid
PubChem CID 71502
IUPAC Name deuterio 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)O

Specifications

Physical Form Liquid
Density 1.50 g/mL
Odor Acrid
Packaging Prescored Ampule
Sensitivity Hygroscopic
Quantity 25 g
Melting Point -15°C
Boiling Point 72°C
UN Number UN2699
Solubility Information Soluble in water.
Formula Weight 115.03
Percent Purity 99.5%
Chemical Name or Material Trifluoroacetic acid-d, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.