missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Non-sterile Transfer Pipettes, Transparent LDPE

Product Code. 13439118 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
4 mL
7 mL
10 mL
Drops/mL:
18
20
25
30
50
Graduated:
No
Yes
Tip Style:
Extended
Extra Long
Jumbo
Standard
Thin Stem
Unit Size:
Pack of 100
Pack of 200
Pack of 400
Pack of 500
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Drops/mL Graduated Tip Style unitSize
13439118 1 mL 30 Yes Extended Pack of 500
13469118 1 mL 30 No Standard Pack of 400
13499108 1 mL 30 Yes Standard Pack of 500
13439108 3 mL 25 Yes Standard Pack of 500
13459118 4 mL 30 No Thin Stem Pack of 500
13459108 4 mL 50 No Standard Pack of 500
13449108 7 mL 20 No Extra Long Pack of 100
13449118 10 mL 18 No Jumbo Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 13439118
Only null left
Add to basket
Edge
Add to basket

Prevent cross contamination. Fisherbrand™ Non-sterile Transfer Pipets, Transparent LDPE are made of shatterproof, flexible plastic and are designed to replace glass Pasteur pipets. Graduated and non-graduated pipets available in a range of sizes and styles, including jumbo, thin stem and extra long.

  • Built-in bulb eliminates the need for plugging, packaging and autoclaving
  • Flexible stem bends to sample from narrow or small tubes
  • Low affinity surface reduces loss of cells and valuable proteins due to binding
  • Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
  • Non-sterile, transparent and fully recyclable

Specifications

Material LDPE
Drops/mL 30
Graduated Yes
Graduations 0.25 mL
Quantity 500 Pack
Tip Style Extended
Volume (Metric) Dispense 33 μL drop
Volume (Metric) 1 mL
Length (Metric) 150 mm
Type Transfer Pipet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.