Learn More
TMEM192 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
| Antigen | TMEM192 |
|---|---|
| Concentration | 367 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16886065
|
Proteintech
28263-1-AP-20UL |
20 μL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16876065
|
Proteintech
28263-1-AP-150UL |
150 μL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TMEM192 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TMEM192. This antibody recognizes human, mouse antigen. The TMEM192 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| TMEM192 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 367 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 201931 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.