missing translation for 'onlineSavingsMsg'
Learn More

Techne™ PrimePro 48 Real-Time PCR System Accessories

Product Code. 15303165
Click to view available options
No. of Wells:
48
Unit Size:
Each
This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.

Product Code. 15303165

Brand: Techne™ 9394598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.

Techne™ PrimePro 48 Real-Time PCR System Accessories keep you running smoothly.

Plates and seals for use with your Techne PrimePro 48 Thermal Cycler

Specifications

Format Standard
Description Real-Time PCR Plates for Prime Pro 48; 50/Pk
Array 6 x 8
Color Natural Translucent
For Use With (Equipment) Techne PrimePro 48
Material Polypropylene
No. of Wells 48
No. per Pack 50
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.