missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE

Product Code. 11963466 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Tip Style:
Beveled
Extended Length, Micro Point, Low Retention
Micro Point
Micro Point, Extended Length
Color:
Clear
Natural
Unit Size:
Pack of 4800
Pack of 960
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Tip Style Color unitSize
11963466 20 to 200 μL Micro Point Natural Pack of 960
11903466 0.1 to 10 μL Micro Point Natural Pack of 960
13418236 0.1 to 10 μL Micro Point, Extended Length Natural Pack of 4800
11913466 0.1 to 10 μL Micro Point, Extended Length Natural Pack of 960
11977714 0.1 to 20 μL Extended Length, Micro Point, Low Retention Clear Pack of 960
13428236 2 to 20 μL Beveled Natural Pack of 4800
11943466 2 to 20 μL Beveled Natural Pack of 960
13438236 10 to 100 μL Beveled Natural Pack of 4800
11953466 10 to 100 μL Beveled Natural Pack of 960
13448236 20 to 200 μL Micro Point Natural Pack of 4800
11987714 30 to 300 μL Micro Point Clear Pack of 960
11973466 100 to 1000 μL Micro Point Natural Pack of 960
13458236 100 to 1000 μL Micro Point Natural Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11963466
Only null left
Add to basket
Add to basket

Choose from a comprehensive line of pipet tips for demanding liquid-handling applications

Universal fit for research grade pipetters.

  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 to 200 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (English) 2 in.
Length (Metric) 58 mm
Format Hinged Rack
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureOne
Graduations 10 μL, 50 μL, 100 μL
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.