Learn More
Memmert™ Support Basket with Perforated Mounting Shelf
Support basket with perforated mounting shelf for conical flasks clips
Brand:  Memmert™ WTB50B48815
 View more versions of this product
Product Code. 17160395
  • ONLINE EXCLUSIVE PRICE
    1865.00 DKK
    1813.01 DKK / Each
    Save 51.99 DKK
    2% Off
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
Description
Description
  • Use of test tube racks not possible
  • Has grid perforations to take clips for conical flasks
Specifications
Specifications
WTB50
Support Basket With Perforated Mounting Shelf (Stainless Steel)
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.