Learn More
Memmert™ Support Basket with Perforated Mounting Shelf
Support basket with perforated mounting shelf for conical flasks clips
Brand:  Memmert™ WTB15B48814
 View more versions of this product
Product Code. 17120315
  • ONLINE EXCLUSIVE PRICE
    943.00 DKK
    917.70 DKK / Each
    Save 25.30 DKK
    2% Off
Only null left
Estimated Shipment: 16-12-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Use of test tube racks not possible
  • Has grid perforations to take clips for conical flasks
Specifications
Specifications
WTB15
Support Basket With Perforated Mounting Shelf (Stainless Steel)
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.