Learn More
Thermo Scientific™ SuperBlock™ Blocking Buffer
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
Brand: Thermo Scientific™ 37545
223.65 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.
Features of SuperBlock Blocking Buffer include:• Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
• Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
• Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

Specifications
SuperBlock (TBS) Blocking Buffer, dry blend, 5 packs | |
5 packs | |
ELISA, Western Blot | |
TBS | |
Upon receipt store at 4°C. Product is shipped at ambient temperature. |
Blocking Buffer, dry blend | |
Powder | |
SuperBlock™ | |
Single purified protein |
Safety and Handling
- SuperBlock Blocking Buffer
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium chloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.