Learn More
Cole-Parmer™ Digital Colony Counter Optional Magnifier
Description
Use this magnifier with Stuart™ SC6 Colony Counters.
- 1.7x and 3x magnifier for CC-200 Colony Counter.
- Pressure sensitive counting.
- Average count facility.
- Bright white energy saving LED lighting.
- With BioCote antimicrobial protection.
- Audible count confirmation.
- Choice of light or dark background.
- Connectivity to printer or computer.
Specifications
Specifications
| For Use With (Equipment) | The Colony Counter Stuart SCPPLUS |
| Includes | BioCote Antimicrobial protection |
| Quantity | 1 Ea. |
| Magnification | 3x |
| Product Type | Lens, Magnifying |