missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences™ Strep-Tactin™ XT Coated Microplate
Shop All IBA Lifesciences Products

Click to view available options
Color:
Clear
Unit Size:
Each
Description
Due to the high specificity and affinity (nM-pM) of the streptavidin variant Strep-Tactin™ XT, Strep-tag™II or Twin-Strep-tag™, fusion proteins can be selectively and stably captured even from complex mixtures. Biomolecules of interest are presented to interacting partners in a uniform manner resulting in reliable and reproducible assays.
- Ready-to-use Strep-Tactin™XT coated microplate (12 x 8-well strips) for ligand binding assays, immunoassays, enzyme assays, or diagnostic assays under high-throughput conditions.
- The solid-phase, multi-well format is compatible with standard multichannel pipettes and automated plate readers.
- Twin-Strep-tag™ fusion protein binding capacity determined with bacterial alkaline phosphatase (97 kDa) is 4 pmol for 200 μL incubation volume.
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Array | 12 x 8 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Material | Plastic |
| Volume (Metric) Well | 300 μL |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Strep-TactinXT |
| Show More |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction