Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
BIOPHARMA
Product Code. 15504004 Shop All Sampling Systems Products
Change view
Click to view available options
Diameter (Metric)Top:
100 mm
200 mm
250 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Sterility unitSize
15504004 200 mm Sterile Pack of 10
16269421 100 mm Sterile Pack of 50
15360044 100 mm Sterile Pack of 100
16259421 100 mm Non-sterile Pack of 50
15879741 100 mm Non-sterile Pack of 100
15889741 200 mm Non-sterile Pack of 20
30286533 250 mm Sterile Pack of 10
30286532 250 mm Non-sterile Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15504004 Supplier Sampling Systems™ Supplier No. 8370H200S
Only null left
Add to basket
Edge
Add to basket
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
  • Constructed of HDPE
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Diameter (Metric)Top 200 mm
Diameter (Metric) Stem 15 mm
Sterility Sterile
Product Type Filling Funnel (Liquid)