missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile IPA
BIOPHARMA

Product Code. 15537171 Shop All Contec Products
Change view
Click to view available options
Container Type:
Capped Bottle
Trigger Spray Bottle
Quantity:
2 x 5 L
6 x 1 L
8 x 0.5 L
Unit Size:
2 bottles
6 bottles
8 bottles
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type Quantity unitSize
15537171 Capped Bottle 2 x 5 L 2 bottles
15892917 Trigger Spray Bottle 8 x 0.5 L 8 bottles
15597171 Trigger Spray Bottle 6 x 1 L 6 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15537171 Supplier Contec™ Supplier No. SBC570I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provided sterile by gamma irradiated using a validated process at no less than 25 kGy. Contec™ Sterile IPA is an alcohol blend of 0.2 micron filtered, filled and bagged in a Grade C (ISO Class 7) cleanroom.

  • Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
  • GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
  • Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
TRUSTED_SUSTAINABILITY

Specifications

Product Type Alcohol
Form Liquid
pH Not Applicable
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Composition Isopropanol in purified water
Concentration 70% (v/v) IPA and 30% Water
Container Type Capped Bottle
Disposable Limited-use
Filtered Filtered
Flammable Yes
For Use With (Application) Sterilizes surfaces
Quantity 2 x 5 L
Volume (English) 1.3 gal.
Volume (Metric) 5 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • CONTEC STERILE 70% IPA 1L AND 5L
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use to extinguish.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.