missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile HydroPure SATWIPES™ System
BIOPHARMA

Contec HydroPure is a blend of 6% hydrogen peroxide with purified water and Amplitude Theta polyester/cellulose wipes.

Brand:  Contec™ SSWC16HP

478.85 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 16341778

  • 687.00€ / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Efficacious against bacteria, fungi, moulds, yeasts and spores, Contec HydroPure leaves little-to-no residue making it ideal for use in critical areas.

Contec HydroPure is provided as a point-of-use SATWIPES system. Each double bagged pack contains a canister of 100 x 54 gsm Amplitude Theta polyester/cellulose dry wipes and a 500 mL capped bottle of Contec HydroPure.

Simply add 500 mL of HydroPure to the dry roll, 5 mins before use, and use the impregnated wipes within 7 days.

Contec HydroPure is 0.2 micron filtered, and filled in a Grade A environment. The sterile product is filtered into pre-irradiated components. The dry wipe canister is irradiated at no less than 25 kGy.

Features & Benefits

  • Impregnated wipes at point-of-use allow convenient and controlled application of disinfectant
  • Hydrogen Peroxide breaks down to water & oxygen leaving no residue
  • Sporicidal activity with very low residue so suitable for critical areas
  • Available sterile so ideal for a Grade A / B areas
  • Double bagged for easy transfer into cleanroom environment
  • Linear tear packaging makes each bag easy to open even when wearing gloves
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dry Wipe
15 cm
23 cm
15 x 23 cm (6 x 9 in.)
100/Canister, 10 Canisters/Cs.
6 in.
9 in.
Polyester Cellulose
Contec HydroPure 6% hydrogen peroxide
6% Hydrogen Peroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CONTEC STERILE SATWIPES HYDROPURE

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide solution

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.