missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ Adipogenesis Differentiation Kit

Product Code. 10154093
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

StemPro™ Adipogenesis Differentiation Kit

For Complete Differentiation of Human Mesenchymal Stem Cells into Fat Cells

StemPro™ Adipogenesis Differentiation Kit has been developed for the adipogenic differentiation of mesenchymal stem cells (MSCs) in tissue culture vessels. The kit contains all reagents required for inducing MSCs to be committed to the adipogenesis pathway and generate adipocytes. Using StemPro™ Adipogenesis Differentiation Kit in combination with StemPro™ MSC SFM or MesenPRO RS™ Medium provides a standardized culture workflow solution for MSC isolation, expansion and differentiation into lipid vesicle-forming adipocytes.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Stem Cells (Mesenchymal)
Content And Storage This kit is shipped in two parts with separate storage requirements. These components are not sold individually:
• StemPro™ Adipocyte Differentiation Basal Medium (100 ml; store in the dark at 2°C to 8°C)
• StemPro™ Adipogenesis Supplement (10 ml; store in the dark at -5°C to -20°C)

The Basal Medium ships at room temperature, while the Supplement ships on dry ice.
Culture Environment CO2
For Use With (Application) Stem Cell Differentiation
Product Type Adipogenesis Differentiation Kit
Endotoxin Level ≤50 EU/mL, ≤ 50 EU/ml
Format Liquid
Glucose High Glucose
Glutamine GlutaMAX™-I
HEPES Buffer No HEPES
Inorganic Salts Calcium, Sodium Phosphate
Phenol Red Indicator No Phenol Red
Product Line Gibco, StemPro
Purity or Quality Grade Research Grade
Quantity 1 Kit
Research Category Differentiation
Serum Level Standard Serum
Sodium Bicarbonate Buffer Sodium Bicarbonate
Sodium Pyruvate Additive Sodium Pyruvate
Species Human
Supplements Adipogenesis Supplements
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.