Learn More
Kinematica Standard Dispersing Aggregate for PT3100 D
Uses two rows of teeth, is suitable for a large spectrum of laboratory applications, and is reliable and efficient.
Brand:  Kinematica 11030298
Product Code. 15515369
  • ONLINE EXCLUSIVE PRICE
    1835.00 €
    1787.00 € / Each
    Save 48.00 €
    2% Off
Only null left
Estimated Shipment: 21-11-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Selection and applications:
    • All standard dispersing tasks
    • Grinding of animal and human tissue samples in a variety of fluids and volumes
    • Sample preparation for the extraction and dissolution of organic materials
    • Sample preparation for medical diagnostic procedures as well as DNA analysis
    • Extractions of systems / substances from vegetable-derived samples
    • Disruption of animal and human cells
    • Deagglomeration of united cell structures
    • Intensive mixing
Specifications
Specifications
Standard Dispersing Aggregate
0.3 to 10 mL
101 mm
1 Ea.
8 m/s
7 mm
PT-DA 07 / 2EC-F101
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.