missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ LabMat™ Bench Liners

Product Code. 11367234 Shop All SP Scienceware Products
Change view
Click to view available options
Dimensions (L x W):
24 x 20 in. (610 x 508mm)
50 ft. x 20 in.
Color:
Orange
White
Description:
One single roll, 50 ft.
Pack of 100 sheets
Unit Size:
1 roll
Each
Pack of 100
Product Code. Dimensions (L x W) Color Description unitSize
11367234 50 ft. x 20 in. White One single roll, 50 ft. Each
11342194 50 ft. x 20 in. Orange One single roll, 50 ft. 1 roll
12918837 24 x 20 in. (610 x 508mm) White Pack of 100 sheets Pack of 100
3 options

Product Code. 11367234

Brand: Bel-Art™ F246750000

Only null left
Add to basket
Edge
Add to basket

Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.

  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials

Specifications

Product Type Bench Protector
Material Polyethylene
Absorbancy 400 mL/m2
Length (English) 50 ft.
Length (Metric) 15.2 m
Width (English) 20 in.
Width (Metric) 508 mm
Dimensions (L x W) 50 ft. x 20 in.
Color White
Description One single roll, 50 ft.
Disposable Single-use
For Use With (Application) Simplify cleanup and safeguard work surfaces from stains, spills and wear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.