Learn More
Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.
Brand:  Bel-Art™ F24675-1000
 View more versions of this product
Product Code. 11342194
  • 106.00 € / 1 roll
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials
Specifications
Specifications
Bench Protector
400 mL/m2
15.2 m
508 mm
Orange
Single-use
Each
Polyethylene
50 ft.
20 in.
50 ft. x 20 in.
One single roll, 50 ft.
Simplify cleanup and safeguard work surfaces from stains, spills and wear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.