Learn More
Syngene™ Dymension 1 Software
Shop All Syngene Products
Click to view available options
For Use With (Equipment):
Gbox-Chemi-XX6/9
Unit Size:
Each
Description
- Software for the rapid analysis of 2D protein spots
between samples containing one gel only - Synchronised zoom with intensity autoscale
- Automatic and precise image warping with vector view
- Background correction and noise filtering
- Automatic spot detection 3 x 10MB images
- Spot detected in eight seconds
- Automatic spot matching
- Spot detection with user adjustable parameters
- Spot editing and filtering Spot normalization
Specifications
Specifications
For Use With (Application) | 2d electrophoresis analysis |
For Use With (Equipment) | Gbox-Chemi-XX6/9 |