missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
HNaO, CAS Number-1310-73-2, sodium hydroxide solution, ascarite, aetznatron, soda lye, sodium hydrate, caustic soda, sodium hydroxide na oh, sodium hydroxide, white caustic, soda, caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 100°C |
| CAS | 1310-73-2 |
| Molecular Formula | HNaO |
| MDL Number | 3548 |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Quantity | 1 L |
| SMILES | [OH-].[Na+] |
| IUPAC Name | sodium;hydroxide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction