missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic)
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
Sodium deuteroxide is a reagent used in the synthesis of rac Zearalenone-d6. is used in many chemical reactions and industrial practices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 10 g |
Assay Percent Range | 99.5% |
Solubility Information | Fully miscible in water. |
Formula Weight | 41.01 |
Odor | Odorless |
Refractive Index | 1.418 |
Sensitivity | Air and moisture sensitive |
Appearance | Colorless liquid |
Show More |
Safety and Handling

Product Identifier
- Sodium deuteroxide, 40% w/w solution in D2O
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Sodium deuteroxide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction