Learn More
Bis(triméthylsilyl)amide de sodium, 98 %, Thermo Scientific Chemicals
Description
Sodium bis(trimethylsilyl)amide is used as a base in organic synthesis as well as to deprotonate other compounds containing weakly acidic O-H, S-H, and N-H bonds. It is used to prepare amines by reacting with alkyl halides. It finds application in certain organic reactions viz. generation of carbenes by dehydrohalogenation of halocarbons and Wittig reagents by deprotonation of phosphonium salts. It accelerates the polymerization of pheneylacetylene in conjunction with rhodium(I) catalysis. Furthermore, it is used for the preparation of lanthanide complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Nom chimique ou matériau | Sodium bis(trimethylsilyl)amide |
| Point de fusion | 171°C to 175°C |
| Couleur | Blanc |
| Odeur | Odorless |
| Formule linéaire | (CH3)3SiNNaSi(CH3)3 |
| Quantité | 2 g |
| Numéro UN | UN3096 |
| Sensibilité | Moisture sensitive |
| Informations sur la solubilité | Soluble in hexane,toluene,ether,terahydrofuran,benzene and toluene. |
| Poids de la formule | 183.38 |
| Afficher plus |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.