missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, Powder, U.S.P. - F.C.C., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
2.5kg
500g
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Formula Weight | 84.01 |
| Grade | FCC/USP |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.20kg |
| Chemical Name or Material | Sodium Bicarbonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction