missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Soda Lime Glass Pasteur Pipets

Product Code. 15212699 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
145 mm
225 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15212699 225 mm Pack of 1000
15202699 145 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15212699 Supplier BRAND™ Supplier No. 747720
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Constructed of Soda Lime Glass and complies with ISO 7712 standards. BRAND™ Soda Lime Glass Pasteur Pipets include a long, drawn-out capillary tip which is perfect for transferring small volumes of aqueous solutions. Suction end with constriction for a cotton plug allows filtering of small quantities.

  • Constructed of Soda Lime Glass.
  • Complies with ISO 7712 standards.
  • Long, drawn-out capillary tip is perfect for transferring small volumes of aqueous solutions.
  • Suction end with constriction for a cotton plug allows filtering of small quantities.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 7712
Diameter (Metric) Outer 7 mm
Material Soda Lime Glass
Quantity 1000 Pack
Tip Style Capillary
Volume (Metric) 2 mL
Length (Metric) 225 mm
Type Pasteur Pipet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.