Learn More
Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes
GREENER_CHOICE
Product Code. 11535564 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 500
Product Code. Color unitSize
11535564 Natural Pack of 500
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11535564 Supplier Thermo Scientific™ Supplier No. 3448
Only null left
Add to basket
Edge
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Improve sample recovery without additives or coatings. These tubes offer maximum sample recovery by significantly reducing surface binding.
Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes improves sample recovery without additives or coatings. The 0.6mL, 1.5mL and 2.0mL snap cap low retention tubes offers maximum sample recovery by significantly reducing surface binding.
  • The duckbill cap reduces the pressure required to open the tube with a wider contact surface yet ensures the cap will remain closed during centrifugation
  • Low retention properties are not achieved by a coating or plastic embedding processes ensuring that samples will not be contaminated from these additives
  • Tubes will remain closed during centrifugation yet open easily with one hand as needed
  • Tubes are available in Amber to protect your light-sensitive samples
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
Color/Packaging Options
  • A variety of packaging options are available to help with contamination control in the lab
  • For stringent sterility requirements, individually wrapped sterile tubes are available
Highest Quality
  • Manufactured from virgin polypropylene these microcentrifuge tubes ensure sample integrity during boiling, freezing, and centrifugation
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Recommended for:
Daily laboratory usage including centrifugation and short term sample storage.
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Flat
Certifications/Compliance Certified free of RNase, DNase, DNA and pyrogens
Description 1.5mL Graduated, Nonsterile
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
Packaging 50 tubes/ bag, 10 bags/ unit, 10 units/ case
Sterility Non-sterile
Graduated Yes
Max. RCF 26,000 x g