Learn More
IKA™ H 100 Silicone Protective Cover
Product Code. 10189972 Shop All IKA Products
Change view
Click to view available options
Type:
Protective Cover
Unit Size:
Each
Product Code. Type unitSize
10189972 Protective Cover Each
1 options
Product Code. 10189972 Supplier IKA™ Supplier No. 0003661000
Only null left
Add to basket
Edge
Add to basket
Use this protective cover with RCT basic magnetic stirrers. IKA™ H 100 Silicone Protective Covers are resistant to most acids, alkaline solutions and organic solvents.
  • Silicone  protective cover
  • For RCT basic s.c. (3380000 + 3810000), included with the magnetic stirrer
  • Max. temperature 135°C
TRUSTED_SUSTAINABILITY
Specifications
Temperature (Metric) 135°C
Type Protective Cover