Learn More
Heidolph™ Boss Head
Product Code. 10124932 Shop All Heidolph Instruments Products
Change view
Click to view available options
Voltage:
115/230 V
Unit Size:
Each
Product Code. Voltage unitSize
10124932 115/230 V Each
1 options
This item is not returnable. View return policy
Product Code. 10124932 Supplier Heidolph™ Supplier No. 5703110000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for stand rods with up to 13.1 mm maximum diameter
Boss head made of anodized aluminum, used to fix retort clamps to stand rods for perfect vertical alignment.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Boss Head
Voltage 115/230 V
Width (Metric) 68
Material Anodized Aluminum
For Use With (Application) Fix retort clamps to stand rods for perfect vertical alignment
Holds Stand rods up to 13.1 mm dia.