Learn More
Grant Instruments™ Liquid McFarland Standards Set
For use with DEN-1 and DEN-1B Densitometers. Grant Instruments™ Liquid McFarland Standards Set is supplied in 16mm dia. glass tubes.
Brand:  Grant Instruments™ DEN MCF STDS
Product Code. 13160623
  • € 504.00 / 5 sets
Please to purchase this item. Need a web account? Register with us today!
Edge
This item is not returnable. View return policy
Description
Description
  • Supplied in 16mm ø glass tubes.
  • 0.5 / 1.0 / 2.0 / 3.0 / 4.0 can be decanted into an alternative tube (shelf-life no longer valid).
  • Requires D16 adaptor, supplied with DEN-1 / DEN-1B
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DEN-1 & DEN-1B Densitometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.