Learn More
Cytiva Sentino™ Microbiology Pump Replacement Tubing Set and Adapter
Product Code. 17114431
Change view
Click to view available options
:
10/Pk.
Product Code.
17114431 10/Pk.
1 options
This item is not returnable. View return policy
Product Code. 17114431 Supplier Cytiva Supplier No. 4861
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Replacement tubing set with MicroFunnel filter funnel adapter.
  • For vacuum filtration of liquids on our Sentino pump
  • For the analysis of aqueous samples for microbiological and particulate contamination using the Membrane Filter (MF) Technique
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MicroFunnel™ Filter Funnel
Sterility Sterile
No. per Pack 10/Pk.