Description
- Safety glasses that self adjust to fit a wide array of head sizes
- 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear to enhance frame comfort for a secure and comfortable fit
- 3M™ Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties, complying with K&N marking requirements of EN166
- Very lightweight, only 18 g
- The flat and flexible temples makes it very popular to use in combination with an Earmuff
Specifications
Specifications
| Certifications/Compliance | CE Approved |
| Lens Coating | Anti-fog |
| Product Type | Safety Spectacles |
| Frame Color | Blue |
| Temple Style | 3M™ Pressure Diffusion Temple Technology |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| UV Protection | UV400 |