missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sanger Sequencing Kit

Product Code. 15869886 Shop All Applied Biosystems Products
Click to view available options
Quantity:
200 Reactions
Unit Size:
Each
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24113” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24113” to get your promotional price

Includes

2 x 100 rxns ExoSAP-IT Express, 2 x BigDye Terminator v1.1 and v3.1 sequencing buffer (5X), 1 x 800 μL BigDye Terminator v3.1 ready reaction mix, 1 tube M13 (-21) primer, 1 tube pGEM control DNA, 1 x 2 mL XTerminator solution, 1 x 9 mL SAM solution

Sanger Sequencing Kit includes all reagents needed for sample preparation for sanger sequencing

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 x 100 rxns ExoSAP-IT Express, store at -5 to -30°C
• 2 x BigDye Terminator v1.1 and v3.1 sequencing buffer (5X), store at 2–8°C
• 1 x 800 µL BigDye Terminator v3.1 ready reaction mix, store at -15 to -25°C
• 1 tube M13 (-21) primer, store at -15 to -25°C
• 1 tube pGEM control DNA, store at -15 to -25°C
• 1 x 2 mL XTerminator solution, store at 2–8°C
• 1 x 9 mL SAM solution, store at room temperature or 4°C

Format Kit
For Use With (Equipment) Sequencing Instruments (Capillary Electrophoresis-Based)
For Use With (Application) Standard Sequencing
No. of Reactions 200
Product Type Sanger Sequencing Kit
Quantity 200 Reactions
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
compliance-icons
Product Identifier
  • Sanger Sequencing Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : benzene

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.