Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
GREENER_CHOICE
Product Code. 15397903
Change view
Click to view available options
Volume (Metric):
20 μL
40 μL
50 μL
60 μL
80 μL
100 μL
150 μL
200 μL
250 μL
300 μL
400 μL
Length (Metric):
5.3 cm
5.9 cm
6.5 cm
7.5 cm
8.0 cm
8.3 cm
8.89 cm
9.5 cm
10.19 cm
10.6 cm
Sterility:
Non-sterile
Description:
100μL Exact Volume Transfer Pipette
150μL Exact Volume Transfer Pipette
20μL Exact Volume Transfer Pipette
200μL Exact Volume Transfer Pipette
300μL Exact Volume Transfer Pipette
40μL Exact Volume Transfer Pipette
400μL Exact Volume Transfer Pipette
50μL Exact Volume Transfer Pipette
60μL Exact Volume Transfer Pipette
80μL Exact Volume Transfer Pipette
Samco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
Unit Size:
Pack of 500
Pack of 5000
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Sterility Description unitSize
15397903 400 μL 10.6 cm Non-sterile 400μL Exact Volume Transfer Pipette Pack of 500
15397803 250 μL 10.19 cm Non-sterile Samco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case) Pack of 500
15595243 20 μL 5.3 cm Non-sterile 20μL Exact Volume Transfer Pipette Pack of 500
15317903 40 μL 5.9 cm Non-sterile 40μL Exact Volume Transfer Pipette Pack of 500
15327903 60 μL 6.5 cm Non-sterile 60μL Exact Volume Transfer Pipette Pack of 500
15176284 80 μL 7.5 cm Non-sterile 80μL Exact Volume Transfer Pipette Pack of 5000
13450778 100 μL 8.3 cm Non-sterile 100μL Exact Volume Transfer Pipette Pack of 5000
15337903 150 μL 8.0 cm Non-sterile 150μL Exact Volume Transfer Pipette Pack of 500
15367903 50 μL 5.9 cm Non-sterile 50μL Exact Volume Transfer Pipette Pack of 500
15377903 200 μL 8.89 cm Non-sterile 200μL Exact Volume Transfer Pipette Pack of 500
15387903 300 μL 9.5 cm Non-sterile 300μL Exact Volume Transfer Pipette Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15397903 Supplier Thermo Scientific™ Supplier No. 967NL
Please to purchase this item. Need a web account? Register with us today!
Edge
Perform precise dispensing of volumes from 20 to 400 μL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
  • Shatterproof, nontoxic polyethylene
  • Nonsterile
  • Low affinity surface
  • Freezer ready
TRUSTED_SUSTAINABILITY
Specifications
Description 400μL Exact Volume Transfer Pipette
Diameter (Metric) Outer 3.8 mm
Material LDPE
Sterility Non-sterile
Graduated No
Packaging Type Bulk 500/Pk 10Pk/Cs 5000/Cs
Volume (Metric) 400 μL
Length (Metric) 10.6 cm
Type Exact Volume Transfer Pipet
Disposable Single-use