missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Richard-Allan Scientific™ Saccomanno Fluid

Product Code. 10046548 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
Unit Size:
14 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10046548 1 gal. 14 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10046548 Supplier Epredia™ Supplier No. 76161

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Store specimens for weeks with Epredia™ Richard-Allan Scientific™ Saccomanno Fluid, which acts as an excellent general cytological fixative.

  • Prevents air drying artifacts
  • For upper respiratory tract cytology, most notably sputum
  • Each prefilled container is labeled with a do not drink warning
  • Clear containers and tinted solution make specimen identification easy
  • Ready-to-use cytology fixative is an alcohol and polyethylene glycol (Carbowax™) solution
TRUSTED_SUSTAINABILITY

Specifications

Capacity 3.79L (1 Gal.)
Product Type Fixative
Quantity 1 gal.
compliance-icons
Product Identifier
  • saccomanno fluid
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.,Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.,Keep container tightly closed.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.,In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.