Learn More
Fisherbrand™ Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector
Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Brand:  Fisherbrand™ 11941-12
 View more versions of this product
Product Code. 15192738
  • 850.00€ / Each
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Compliance
WEEE and ROHS compliant
Specifications
Specifications
Deuterium
0.16 kg
1100 DAD, 8453 HPLC Detector
0.36 lb.
Item 14-500-721 is comparable to Agilent part number 2140-0605.
Detector Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.