Learn More
Corning™ Regular Fetal Bovine Serum
BIOPHARMAGREENER_CHOICE
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Brand:  Corning™ 35-010-CV
 View more versions of this product
Product Code. 15343681
  • POA / 500mL
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)
Order Info
Shipped in frozen form
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cGMP
-25° to -15°C
500 mL
Triple-filtered, 0.1 μm
Regular
Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
<30 mg/dL
7.6 ±0.8
USDA Inspected
Amber Yellow to Reddish
Fetal Bovine Serum
Sterile
Bovine Fetus
≤20EU/ mL
No
315 ± 35mOsm/kg
30 to 50g/L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For research use only.