missing translation for 'onlineSavingsMsg'
Learn More

Redox Buffer, 220mV, Reagecon™
BIOPHARMA

Product Code. 11736398 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. Quantity unitSize
11736398 250 mL 250mL
1 options
This item is not returnable. View return policy

Product Code. 11736398

Brand: Reagecon 90565

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for testing, calibration and verification of redox/ORP sensors. Redox Buffer, 220mV, Reagecon™ is a ready to use solution of iron salts, including potassium ferrocyanide and tripotassium hexacyanoferrate, for use with reactions with a redox potential of 220mV.

Redox (reduction–oxidation) reactions cause an electron transfer process that can be measured using a Redox or ORP (oxidation-reduction potential) electrode. Redox is an absolute measurement expressed in millivolts. During its working life, a Redox electrode undergoes no change of zero point or slope. Therefore, redox electrodes do not require calibration. A control standard controls the functionality of the sensing and reference electrode, and also controls the analyst's technique, environmental conditions and the operation of the measurement meter.

Specifications

CAS 13943-58-3, 13746-66-2
Melting Point 0°C
Boiling Point 100°C
For Use With (Application) Test ORP electrodes
For Use With (Equipment) Redox and ORP Sensors
Physical Form Liquid
Quantity 250 mL
Formulation Iron salts
Odor Characteristic
Vapor Pressure 23 hPa at 20°C
Solubility Information Fully miscible in water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.