missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Reagent Reservoir, Sterile Polystyrene
BIOPHARMAGREENER_CHOICE

Product Code. 16354852 Shop All Thermo Scientific Incubateurs à CO2 Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
Packaging:
1/bag, 100 bags
1/bag, 50 bags
5/bag, 20 bags
5/bag, 40 bags
Dimensions (L x W x H):
2.8 x 0.8 x 0.7 in
4 x 1 x 0.8 in
4 x 1.1 x 1.2 in
4.3 x 1.9 x 1.3 in
Unit Size:
Pack of 100
Pack of 200
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Packaging Dimensions (L x W x H) unitSize
16354852 10 mL 1/bag, 100 bags 2.8 x 0.8 x 0.7 in Pack of 100
16334852 5 mL 1/bag, 50 bags 2.8 x 0.8 x 0.7 in Pack of 50
16364852 25 mL 5/bag, 40 bags 4 x 1 x 0.8 in Pack of 200
16374852 25 mL 1/bag, 50 bags 4 x 1 x 0.8 in Pack of 100
16314862 50 mL 5/bag, 20 bags 4 x 1.1 x 1.2 in Pack of 100
16394852 50 mL 5/bag, 40 bags 4 x 1.1 x 1.2 in Pack of 200
16317701 50 mL 1/bag, 100 bags 4 x 1.1 x 1.2 in Pack of 100
16324862 100 mL 5/bag, 40 bags 4.3 x 1.9 x 1.3 in Pack of 200
16334862 100 mL 1/bag, 100 bags 4.3 x 1.9 x 1.3 in Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 16354852 Supplier Heathrow Scientific™ Supplier No. 120639
Only null left
Add to basket
Edge
Add to basket

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures

  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents
  • Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Quality certificate available on request
  • Disposable
  • Design Patent Pending

Specifications

Capacity (English) .34 oz
Certifications/Compliance Dnase, Rnase & Pyrogen Free
Coating None
Color White
Description Single wrap
Capacity (Metric) 10 mL
Dimensions (L x W x H) 2.8 x 0.8 x 0.7 in
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) Reagent
Height (English) 0.7 in
Height (Metric) 1.8 cm
Includes Reservoir
Length (English) 2.8 in
Length (Metric) 7.2 cm
Max. Temperature (English) 104°F Operating
Max. Temperature (Metric) 40°C Operating
No. per Case 600
No. per Pack 100
Packaging 1/bag, 100 bags
Volume (Metric) Well 10 mL
Width (English) 0.8 in
Width (Metric) 2.1 cm
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Conical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.