missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Quiltec™ I Mop Head
BIOPHARMA

Product Code. 15817044 Shop All Contec Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
15817044 Sterile Pack of 40
15815597 Non-sterile Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15817044 Supplier Contec™ Supplier No. MHQT0150
Only null left
Add to basket
Edge
Add to basket

Contec Quiltec I Mop Heads are manufactured from 100% 2-ply polyester around a rayon core so are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.

  • Two-ply 100% knitted polyester over a rayon core
  • Gamma irradiated at no less than 25 kGy
  • 5 mops double bagged
  • Approx 18-20 sqm coverage
  • Certificates of conformity supplied with every batch
TRUSTED_SUSTAINABILITY

Specifications

Dimensions 39 x 13 cm
Product Type Mop Head
Color Code White
Sterility Sterile
Autoclavable Not Autoclavable
Disposable Single-use
For Use With (Application) Cleanroom Mopping
Color-coded No
Material Polyester, Rayon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.