Learn More
Macherey-Nagel™ Quantofix™ Nickel Test Strip Kits
Allows for the quick and easy determination of nickel in solutions. Macherey Nagel™ Quantofix™ Nickel Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Brand:  Macherey-Nagel™ 91305
Product Code. 10100491
  • 34.15 € / Pack of 100
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Allows for the quick and easy determination of nickel in solutions. Macherey Nagel™ Quantofix™ Nickel Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Ready to use
  • Rapid: provides reliable results within 30 seconds
  • Excellent for use in metal plating processes and electroplating solutions
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C
Specifications
Specifications
White to Bright Red
Determination of Nickel
100 Pack
30 sec.
Strip
0, 10, 25, 50, 100, 250, 500, 1000mg/L Ni2+
30°C
Semi-qualitative Test Strip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.