missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d{5}, 99.5% (Isotopic), contains 0.05% v/v TMS

Product Code. 11334438
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7291-22-7 | C5H5N | 84.13 g/mol

Pyridine-d{5} is used in dehalogenation reactions and can be used as a base in condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7291-22-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 84.13
MDL Number MFCD00044639
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
PubChem CID 558519
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

Specifications

Physical Form Liquid
Density 1.05 g/mL
Flash Point 20°C (68°F)
Odor Unpleasant
Refractive Index 1.506
Quantity 5 g
Melting Point -42°C
Boiling Point 115°C
UN Number UN1282
Beilstein 114377
Solubility Information Soluble in water,alcohol,ether,acetone,and benzene.
Formula Weight 84.14
Percent Purity 99.5%
Concentration or Composition (by Analyte or Components) Contains 0.05% v/v TMS
Chemical Name or Material Pyridine-d5, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.