missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ 60-layer EnviroTack™ Contamination Control Mats
BIOPHARMAGREENER_CHOICE

Product Code. 15371992 Shop All Purus Products
Change view
Click to view available options
Color:
Blue
Gray
White
Dimensions (L x W):
18 x 36 in.
18 x 45 in.
24 x 30 in.
24 x 36 in.
26 x 45 in.
36 x 36 in.
36 x 45 in.
Surface Coating:
High Tack
Regular Tack
Unit Size:
Pack of 4
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Surface Coating unitSize
15371992 Gray 36 x 45 in. Regular Tack Pack of 4
15311982 Gray 26 x 45 in. Regular Tack Pack of 4
15763619 White 24 x 30 in. Regular Tack Pack of 4
15361992 Blue 36 x 45 in. Regular Tack Pack of 4
15311962 Blue 18 x 36 in. Regular Tack Pack of 4
15321962 Gray 18 x 36 in. Regular Tack Pack of 4
15371962 Blue 18 x 45 in. Regular Tack Pack of 4
15381962 Gray 18 x 45 in. Regular Tack Pack of 4
15743619 Blue 24 x 30 in. Regular Tack Pack of 4
15753619 Gray 24 x 30 in. Regular Tack Pack of 4
15391962 White 18 x 45 in. Regular Tack Pack of 4
15331972 Blue 24 x 36 in. Regular Tack Pack of 4
15341972 Gray 24 x 36 in. Regular Tack Pack of 4
15351972 White 24 x 36 in. Regular Tack Pack of 4
15381972 Blue 26 x 45 in. Regular Tack Pack of 4
15321982 White 26 x 45 in. Regular Tack Pack of 4
15381982 Blue 36 x 36 in. Regular Tack Pack of 4
15331962 White 18 x 36 in. Regular Tack Pack of 4
15391992 White 36 x 45 in. High Tack Pack of 4
15381992 White 36 x 45 in. Regular Tack Pack of 4
15391982 Gray 36 x 36 in. Regular Tack Pack of 4
15391972 Blue 26 x 45 in. High Tack Pack of 4
15331982 White 26 x 45 in. High Tack Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15371992 missing translation for 'mfr' PURUS™ missing translation for 'supplierNo' PS 3645 64 G
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels

  • Eco-friendly containing a proprietary GMO free, plant based renewable material that reduces CO2 emissions during the manufacturing process
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams or through Fisher Scientific's partnership with PURUS™ and Terra Cycle
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers

Certifications

Manufactured in ISO 9001 certified facility

Specifications

Certifications/Compliance Manufactured in an ISO 9001 certified facility
Dimensions (L x W) 36 x 45 in.
For Use With (Application) For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
Quantity 60 Sheets/Mat, 4 Mats/Cs.
Surface Coating Regular Tack
Thickness (English) Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil
Thickness (Metric) Film: 33 μm Adhesive: 8 μm
Color Gray
Material LDPE Blend
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.