Learn More
uvex Pure-Fit Safety Glasses
GREENER_CHOICE
Product Code. 17533085 Shop All Uvex Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Uncoated
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating unitSize
17533085 Anti-fog, Scratch-resistant Each
17523085 Uncoated Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17533085 Supplier uvex Supplier No. 9145265
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Glasses designed to be recycled. Pure-Fit can be manufactured fully automatically from just one material without the need for additional assembly.
  • Reduced to the max - pure, ultralight safety spectacle with innovative side arm concept weighing only 19 grams
  • Frameless lens design for a wide field of vision
  • X-tended sideshield offers additional protection from hazards
  • Flexible side arms deliver the perfect secure, pressure-free fit
  • Nose bridge designed for all-day comfort
  • Robust polycarbonate lens
  • UV400 – 100 % protection against dangerous UVA and UVB radiation up to 400 nm
  • Metal-free
  • Frame transparent / W 166 FT CE
  • Lens PC clear, UV400 / 2C-1,2 W1 FT CE
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Frame Color Transparent
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Standard