Learn More
Contec™ PROSAT™ Sterile Low Endotoxin Wipes



Made from 100% knitted polyester Polynit Heatseal and presaturated with a blend of 70% IPA and the highest grade distilled Water for Injection (WFI). PROSAT™ Sterile Low Endotoxin Wipes are low in particles and fibers for ideal usage in product contact areas. Each batch of wipes has a guaranteed endotoxin level of less than <1EU per device.
Brand: Contec™ PSWE0003-BPR
7172.78 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- PROSAT™ Sterile™ Polynit Heatseal LE Wipes are made of 100% knitted polyester Polynit Heatseal, presaturated with a blend of 70% isopropanol and 30% water for injection (WFI).
- Each lot is tested before release and low endotoxin certified to less than <1EU/wipe.
- The sealed edge wipe produces very low levels of particles and fibers and is ideal for use on product contact surfaces.
- Each resealable pouch contains a small number of wipes, eliminating any waste at the end of a session.
- Each pouch is individually bagged for easy entry into the sterile suite.
- Presaturated wipes provide many benefits including solvent control, reduced Volatile Organic Compounds (VOCs) and increased process control and repeatability.
- The resealable pouch reduces waste.
- PROSAT Sterile Polynit Heatseal LE Wipes are validated sterile and suitable for use in aseptic suites and the most critical pharmaceutical cleanrooms.
Features and Benefits:
- Recommended for Grade A/B or ISO Class 3-8 environments - product contact areas
- 100% knitted polyester fabric results in very low levels of particles and fibers
- Laser sealed edges reduces fiber generation from the edge of the wipe
- Guaranteed endotoxin levels of <1EU/device make them ideal for product contact surfaces
- Small number of wipes per resealable pouch to eliminate waste
- Small footprint, hanging pouches available, so ideal for use in isolator and RAB systems
- Presaturated wipe reduces VOC emissions and solvent use
- Resealable pouch preserves cleanliness and solvent saturation levels

Specifications
Pre-saturated Cleanroom Wipe | |
30 cm | |
30 cm | |
Not Autoclavable | |
No-run interlock knit | |
30 x 30 cm (12 x 12 in.) | |
10 Wipes/Pouch, Triple-Bagged | |
70% IPA/ 30% WFI, USP grade |
12 in. | |
12 in. | |
Polyester | |
140 gsm | |
Single-use | |
Most Critical Pharmaceutical Cleanrooms | |
2-ply | |
Sterile |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.