missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Presaturated Nonwoven Wipes, Quarterfold
BIOPHARMA

Product Code. p-9514001 Shop All Contec Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 600
Pack of 750
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
17204523 Non-sterile Pack of 750
17214523 Sterile Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17204523 Supplier Contec™ Supplier No. PSLPP7030BPR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.

  • Wipe substrate designed for the application
  • Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping articles prior to pass through
  • Made of meltblown polypropylene or cellulose/polyester
  • Effective and convenient for wipe-downs of process tools and other equipment
  • Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
  • Solvent blend meets SEMI specifications for metals and contaminants
TRUSTED_SUSTAINABILITY

Specifications

Product Type Disinfectant Wipes
Length (English) 17 in.
Length (Metric) 43 cm
Width (English) 16 in.
Width (Metric) 40.6 cm
Material Polypropylene
Autoclavable Not Autoclavable
Certifications/Compliance ISO Class 5, Authorized for compliance with the requirements of the EU Biocides Regulation (BPR)
Disposable Single-use
Dimensions (L x W) 43 x 41 cm (17 x 16 in.)
For Use With (Application) For interior surfaces of process tools, surface preparation, manufacturing processes
For Use With (Equipment) Process tools and other equipment
Packaging Quantity 50 Wipes/Bag
Ply 1-ply
Saturation Solution 70% IPA/ 30% DI Water
Sterility Non-sterile
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.