Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 2%
Descrizione
Includes
E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, SYBR Safe 2%, provides everything you need to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the device.
The kit includes:
- 18 E-Gel agarose gels with SYBR Safe, 2% (Cat. No. G521802)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
Warranty
1 year warranty
Specifica
Specifica
| Content And Storage | • 1 electrophoresis device • 18 E-Gel SYBR Safe 2% gels • 1 vial E-Gel 50 bp DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter Store at room temperature. |
| Gel Size | Mini |
| Tipo | Electrophoresis Device Starter Kit |
| Shipping Condition | Room Temperature |
| Tensione max | 240 V AC (Power Supply), 48 V DC (Electrophoresis Device) |
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.